Learn More
MSC 80/20 Inc. Hidden Corner Connector, Inside-Outside, 45 Series, Zinc
Catalog No. 5000532332
Encompass
This hidden corner connector provides a 90 degree hidden corner connection. It includes set screws for quick and easy connections. It allows for connections between the inside of two profiles or between the inside of one profile and the outside of a second profile.
  • Provides a 90-degree hidden corner connection
  • Comes with set screws for quick and easy connections
  • Allows connection between the inside of two profiles or between the inside of one profile and the outside of the second profile
  • Does not require any machining
  • Cost-effective fastening option
  • Suggested tool to tighten included hardware: 4 mm hex wrench
Catalog No. 50-005-32332 Supplier MSC Supplier No. 32059776
This hidden corner connector provides a 90 degree hidden corner connection. It includes set screws for quick and easy connections. It allows for connections between the inside of two profiles or between the inside of one profile and the outside of a second profile.
  • Provides a 90-degree hidden corner connection
  • Comes with set screws for quick and easy connections
  • Allows connection between the inside of two profiles or between the inside of one profile and the outside of the second profile
  • Does not require any machining
  • Cost-effective fastening option
  • Suggested tool to tighten included hardware: 4 mm hex wrench