Learn More
MSC 80/20 Inc. Hidden Corner Connector, Inside-to-Inside, for Series 45 Series, Zinc
Catalog No. 5000533075
Encompass
This inside-to-inside hidden corner connector is designed for creating 90-degree connections within two profiles. Made from durable zinc, it is specifically for use with Series 45 Series profiles, providing a concealed and robust structural link.
  • Designed for inside of two profiles
  • Provides a hidden corner connection
  • Forms a 90-degree connection
  • Constructed from zinc for durability
Catalog No. 50-005-33075 Supplier MSC Supplier No. 32059784
This inside-to-inside hidden corner connector is designed for creating 90-degree connections within two profiles. Made from durable zinc, it is specifically for use with Series 45 Series profiles, providing a concealed and robust structural link.
  • Designed for inside of two profiles
  • Provides a hidden corner connection
  • Forms a 90-degree connection
  • Constructed from zinc for durability