Learn More
MSC 80/20 Inc. Hidden Corner Connector, Inside-to-Outside, Series 30/40, Zinc, 30 mm
Catalog No. 5000527709
Encompass
This Inside-to-Outside Hidden Corner Connector is designed for use with Series 30/40 profiles. Made from zinc, it offers a secure and cost-effective fastening solution. It has an overall length of 30 mm and features an M6 thread size.
  • Inside-to-outside hidden corner connector
  • Designed for use with Series 30/40 profiles
  • Constructed from zinc for durability
  • Features a clear finish
  • Overall length of 30 mm
  • Includes a 3mm hex wrench for installation
  • Offers a cost-effective fastening option
  • Equipped with an M6 thread size
Catalog No. 50-005-27709 Supplier MSC Supplier No. 69451136
This Inside-to-Outside Hidden Corner Connector is designed for use with Series 30/40 profiles. Made from zinc, it offers a secure and cost-effective fastening solution. It has an overall length of 30 mm and features an M6 thread size.
  • Inside-to-outside hidden corner connector
  • Designed for use with Series 30/40 profiles
  • Constructed from zinc for durability
  • Features a clear finish
  • Overall length of 30 mm
  • Includes a 3mm hex wrench for installation
  • Offers a cost-effective fastening option
  • Equipped with an M6 thread size