Learn More
MSC 80/20 Inc. Inside Corner Bracket, 40 Series, 2 Hole, Aluminum, Clear Anodized
Supplier:  MSC 05224225
Encompass
This inside corner bracket is designed to create a 90-degree connection in T-slot aluminum extrusions. Made from clear anodized aluminum, it offers infinite positioning along the profile T-slot without requiring machining services, ensuring a high degree of adjustability. The bracket engages a 2-degree drop-lock feature when assembly bolts are tightened, and the fastening method is both repositionable and reusable.
  • Creates a 90-degree connection
  • Requires no machining services
  • Provides infinite positioning along the profile T-slot
  • Engages a 2-degree drop-lock feature
  • Repositionable and reusable
  • Constructed from aluminum with a clear anodized finish
Catalog No. 50-005-29724