Learn More
MSC 80/20 Inc. Linear Bearing, 25 Series, 4 Slot Mount, Double Flange, Short Standard
Catalog No. 5000532839
Encompass
This linear bearing is designed for linear motion applications, specifically for housing profiles with a minimum of four open T-slots. It features a 4-slot mount, a double flange, and a short standard length, making it suitable for various industrial uses. The bearing is supplied with self-lubricating UHMW pads, screws, and shims.
  • Designed for compression loads only.
  • Not intended to support hanging loads.
  • Constructed from aluminum grade 6063-T6.
  • Features an extruded process and clear anodize finish.
  • Compatible with 25 series profiles.
  • Includes self-lubricating UHMW pads, screws, and shims.
Catalog No. 50-005-32839 Supplier MSC Supplier No. 54969670
This linear bearing is designed for linear motion applications, specifically for housing profiles with a minimum of four open T-slots. It features a 4-slot mount, a double flange, and a short standard length, making it suitable for various industrial uses. The bearing is supplied with self-lubricating UHMW pads, screws, and shims.
  • Designed for compression loads only.
  • Not intended to support hanging loads.
  • Constructed from aluminum grade 6063-T6.
  • Features an extruded process and clear anodize finish.
  • Compatible with 25 series profiles.
  • Includes self-lubricating UHMW pads, screws, and shims.