Learn More
MSC 80/20 Inc. Linear Bearing, Double Flange, 15 Series, Aluminum, 5.5 in
Catalog No. 5000516843
Encompass
This double flange linear bearing is designed for linear motion applications. It features a 4-slot mount and houses profiles with a minimum of four open T-slots. The bearing includes self-lubricating UHMW pads, screws, and shims, and is intended for compression loads only.
  • Double flange design
  • 4-slot mount for versatile installation
  • Longer option for extended applications
  • Houses profiles with a minimum of four open T-slots
  • Includes self-lubricating UHMW pads, screws, and shims
  • Constructed from aluminum with an anodized finish
  • Compatible with 15 Series extrusions
  • Designed for compression loads in linear motion applications
Catalog No. 50-005-16843 Supplier MSC Supplier No. 07172828
This double flange linear bearing is designed for linear motion applications. It features a 4-slot mount and houses profiles with a minimum of four open T-slots. The bearing includes self-lubricating UHMW pads, screws, and shims, and is intended for compression loads only.
  • Double flange design
  • 4-slot mount for versatile installation
  • Longer option for extended applications
  • Houses profiles with a minimum of four open T-slots
  • Includes self-lubricating UHMW pads, screws, and shims
  • Constructed from aluminum with an anodized finish
  • Compatible with 15 Series extrusions
  • Designed for compression loads in linear motion applications