Learn More
MSC 80/20 Inc. Roller Wheel, 15 Series, Dual Roller Bearing, Nylon, Fiberglass Reinforced
Catalog No. 5000529534
Encompass
This roller wheel features a dual roller bearing for smooth movement within profile T-slots. Constructed from fiberglass-reinforced nylon, it is designed for moving frames, panels, and components efficiently.
  • Constructed from nylon PA glass fiber reinforced grade K222-D.
  • Features a dual roller bearing.
  • Injection molded for durability.
  • Allows for smooth movement within profile T-slots.
  • Designed for moving frames, panels, and components.
  • Compatible with 15 series applications.
Catalog No. 50-005-29534 Supplier MSC Supplier No. 07217631
This roller wheel features a dual roller bearing for smooth movement within profile T-slots. Constructed from fiberglass-reinforced nylon, it is designed for moving frames, panels, and components efficiently.
  • Constructed from nylon PA glass fiber reinforced grade K222-D.
  • Features a dual roller bearing.
  • Injection molded for durability.
  • Allows for smooth movement within profile T-slots.
  • Designed for moving frames, panels, and components.
  • Compatible with 15 series applications.