Learn More
MSC 80/20 Inc. Roller Wheel, Permanent Lubricated Bushing, Nylon PA, Black
Catalog No. 5000529730
Encompass
This roller wheel is engineered for guided linear motion applications. It features a permanent lubricated bronze bushing, removing the need for external lubrication. Constructed from durable Nylon PA glass fiber reinforced material, this component is black and designed for easy mounting to profile ends or T-slots.
  • Engineered for guided linear motion
  • Bronze bushing eliminates the need for additional lubrication
  • Mounts to the profile end or T-slot
  • Includes mounting hardware
  • Constructed from Nylon PA glass fiber reinforced material
  • Black color
  • Compatible with 10 series systems
  • Injection molded for precision
Catalog No. 50-005-29730 Supplier MSC Supplier No. 06036388
This roller wheel is engineered for guided linear motion applications. It features a permanent lubricated bronze bushing, removing the need for external lubrication. Constructed from durable Nylon PA glass fiber reinforced material, this component is black and designed for easy mounting to profile ends or T-slots.
  • Engineered for guided linear motion
  • Bronze bushing eliminates the need for additional lubrication
  • Mounts to the profile end or T-slot
  • Includes mounting hardware
  • Constructed from Nylon PA glass fiber reinforced material
  • Black color
  • Compatible with 10 series systems
  • Injection molded for precision