Learn More
MSC 80/20 Inc. Structural Pivot Assembly, 15 Series, 0 Degree, Dual Straight Arms, Aluminum
Catalog No. 5000514196
Encompass
This standard structural pivot assembly with dual straight arms comes preassembled. It includes the pivot arms connected to the nub and all necessary hardware to connect the nub to the mounting profile. The assembly consists of one 0 degree, 90 degree or universal structural pivot nub and two straight structural pivot arms, offering versatile configuration options.
  • Compatible with 15 Series profiles
  • Constructed from aluminum
  • Precision milled with 6063-T6 grade aluminum
  • Features an anodized finish
  • Designed for use with Bolt Kit 3320 or 3325
  • Includes hardware for arm to nub and nub to T-slot connection
Catalog No. 50-005-14196 Supplier MSC Supplier No. 76824515
This standard structural pivot assembly with dual straight arms comes preassembled. It includes the pivot arms connected to the nub and all necessary hardware to connect the nub to the mounting profile. The assembly consists of one 0 degree, 90 degree or universal structural pivot nub and two straight structural pivot arms, offering versatile configuration options.
  • Compatible with 15 Series profiles
  • Constructed from aluminum
  • Precision milled with 6063-T6 grade aluminum
  • Features an anodized finish
  • Designed for use with Bolt Kit 3320 or 3325
  • Includes hardware for arm to nub and nub to T-slot connection