Learn More
MSC 80/20 Inc. T-Slot Framing, Aluminum Alloy, Standard Duty, 3 Meter
Catalog No. 5000511419
Encompass
This T-Slot Framing from 80/20 Inc. is designed for various construction applications, offering a robust and versatile solution. Crafted from durable aluminum alloy, it provides a standard-duty option for creating custom frames, enclosures, and workstations.
Features and Benefits:
  • T-Slotted frame type for versatile assembly
  • Standard-duty construction for reliable performance
  • Manufactured from high-quality aluminum alloy
  • Length of 3 meters for extensive project requirements
Catalog No. 50-005-11419 Supplier MSC Supplier No. 05269238
This T-Slot Framing from 80/20 Inc. is designed for various construction applications, offering a robust and versatile solution. Crafted from durable aluminum alloy, it provides a standard-duty option for creating custom frames, enclosures, and workstations.
Features and Benefits:
  • T-Slotted frame type for versatile assembly
  • Standard-duty construction for reliable performance
  • Manufactured from high-quality aluminum alloy
  • Length of 3 meters for extensive project requirements