Learn More
MSC 80/20 Inc. T-Slotted Aluminum Extrusion, 1.50 x 1.50 in, Anodized Black, 72 in
Catalog No. 5000537401
Encompass
This T-slotted aluminum extrusion features a 1.50 x 1.50 in lite fractional square profile with four open T-slots, one on each 1.50 in face. It includes align-a-grooves for alignment assistance and is compatible with all 15 series fasteners. This profile is suitable for applications such as machine guards, enclosures, or workbenches, providing multi-directional access for mounting accessories.
  • Features a 1.50 x 1.50 in lite fractional square T-slot profile
  • Equipped with four open T-slots for versatile accessory mounting
  • Includes align-a-grooves for simplified alignment during connection
  • Compatible with all 15 series fasteners for broad utility
  • Constructed from durable aluminum with an anodized black finish
  • Ideal for building machine guards, enclosures, and workbenches
  • Measures 72 in in height for various framing applications
Catalog No. 50-005-37401 Supplier MSC Supplier No. 61998597
This T-slotted aluminum extrusion features a 1.50 x 1.50 in lite fractional square profile with four open T-slots, one on each 1.50 in face. It includes align-a-grooves for alignment assistance and is compatible with all 15 series fasteners. This profile is suitable for applications such as machine guards, enclosures, or workbenches, providing multi-directional access for mounting accessories.
  • Features a 1.50 x 1.50 in lite fractional square T-slot profile
  • Equipped with four open T-slots for versatile accessory mounting
  • Includes align-a-grooves for simplified alignment during connection
  • Compatible with all 15 series fasteners for broad utility
  • Constructed from durable aluminum with an anodized black finish
  • Ideal for building machine guards, enclosures, and workbenches
  • Measures 72 in in height for various framing applications