Learn More
MSC 80/20 Inc. T-Slotted Extrusion Support Bracket, 45 Degree, Aluminum, 12 Inch
Catalog No. 5000534482
Encompass
This T-Slotted Extrusion Support Bracket is designed for use with Series 10 - 1010 Extrusion systems. Made from durable aluminum, this 45-degree support assembly, measuring 12 inches overall length, helps to minimize beam deflection and ensures a flush connection for your structural applications.
  • Designed for series 10 - 1010 extrusion.
  • Constructed from aluminum.
  • Features a 45-degree support assembly.
  • Includes necessary washers and T-nuts.
  • Minimizes beam deflection.
  • Provides a flush connection.
  • Measures 12 inches overall length, 1 inch deep, and 12 inches high.
  • Thread size is 1/4-20.
Catalog No. 50-005-34482 Supplier MSC Supplier No. 73097388
This T-Slotted Extrusion Support Bracket is designed for use with Series 10 - 1010 Extrusion systems. Made from durable aluminum, this 45-degree support assembly, measuring 12 inches overall length, helps to minimize beam deflection and ensures a flush connection for your structural applications.
  • Designed for series 10 - 1010 extrusion.
  • Constructed from aluminum.
  • Features a 45-degree support assembly.
  • Includes necessary washers and T-nuts.
  • Minimizes beam deflection.
  • Provides a flush connection.
  • Measures 12 inches overall length, 1 inch deep, and 12 inches high.
  • Thread size is 1/4-20.