Learn More
MSC 80/20 Inside Corner Bracket, 10 Series, 8 Hole, Anodized Aluminum 6063-T6
Catalog No. 5000530303
Encompass
This 10 Series 8 Hole Inside Corner Bracket is an external fastening method designed to create robust 90-degree connections for structural framing. Made from anodized 6063-T6 aluminum, it provides infinite positioning along the profile T-slot, allowing a large degree of adjustability.
  • External fastening for straightforward installation.
  • Creates a 90-degree connection.
  • Features four holes on each side, arranged in two rows and two columns.
  • Requires no machining services.
  • Offers infinite positioning along the profile T-slot, providing significant adjustability.
  • Engages a 2-degree drop-lock feature when assembly bolts are tightened.
  • Repositionable and reusable for various applications.
Catalog No. 50-005-30303 Supplier MSC Supplier No. 71825020
This 10 Series 8 Hole Inside Corner Bracket is an external fastening method designed to create robust 90-degree connections for structural framing. Made from anodized 6063-T6 aluminum, it provides infinite positioning along the profile T-slot, allowing a large degree of adjustability.
  • External fastening for straightforward installation.
  • Creates a 90-degree connection.
  • Features four holes on each side, arranged in two rows and two columns.
  • Requires no machining services.
  • Offers infinite positioning along the profile T-slot, providing significant adjustability.
  • Engages a 2-degree drop-lock feature when assembly bolts are tightened.
  • Repositionable and reusable for various applications.