Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

80% Ethanol, Molecular Biology Grade

Catalog No. T08204K7 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T08204K7 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T08204K7
 Restricted Item
Add to Cart

missing translation for 'validMainframeMsg'

Ready made ethanol/water blend prequalified for molecular biology research applications. | CAS: 64-17-5 | C2H6O | 46.07 g/mol

  • 80% ethanol and water blend
  • DNase-, RNase-, and protease-free
  • Ready made ethanol/water blend
  • Pre-qualified for molecular biology research applications
  • Suitable for the purification and precipitation of nucleic acids (DNA and RNA) and proteins
  • Tested for use with Applied Biosystems™ MagMAX™ kits
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Color Clear, Colorless Liquid
Quantity 4 L
Packaging Poly Bottle
Residue after Evaporation 0.001% Maximum
Assay Percent Range 78% to 82%
Percent Purity 80%
Grade Molecular Biology
Chemical Name or Material Ethyl Alcohol
Hazard Category Serious eye damage/eye irritation (category 2), Flammable liquids (category 2)
Hazard Statement Causes serious eye irritation, Highly flammable liquid and vapor

For Manufacturing and Laboratory Use. Not for beverage use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.