Learn More
ELKO FILTERING CO 80 MICRON MESH 1YARD
SDP
Catalog No. NC0422724
Encompass
Nylon Monofilament Precision Woven Mesh - 80 Micron Opening, Open Area 37%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-80/37
Catalog No. NC0422724 Supplier ELKO FILTERING CO Supplier No. 0380/37
Add to Cart
Add to Cart
Nylon Monofilament Precision Woven Mesh - 80 Micron Opening, Open Area 37%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-80/37