Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

80 Octane Blend, PRF, Meets requirements of ASTM D2699 and D2700, Solstice

Catalog No. 6000184 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Steel Can
Steel Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-001-84 204 L Steel Drum
60-001-83 19 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-001-84 Supplier Solstice Supplier No. RS2175204
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material 80 Octane Blend, PRF
Name Note B&J Brand, Meets requirements of ASTM D2699 and D2700
CAS 540-84-1, 142-82-5
Boiling Point 100°C
CAS Max % 80.0000%, 20.0000%
UN Number UN1993
Density 0.6859 g/cm3
Grade Primary Reference Fuel
Packaging Steel Drum
Quantity 204 L
Flash Point −12.2°C
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.