Learn More
Andwin Scientific 80-PLACE TUBE RACKS FOR 0.5 1
SDP
Catalog No. 50819978
Encompass_Preferred
Made with polypropylene, the tube rack can store up to 80 1.5 ml and 2.0 ml tubes, including its screw cap tubes. The rack can conveniently endure autoclaving and freezing temperatures of up to (-90C). This product measures 9" x 2 5/8" x 1 1/8" and is sold in a pack of 7 racks in assorted colors of blue, green, orange, red, yellow, violet, and natural.
This item is not returnable. View return policy
Catalog No. 50-819-978 Supplier Andwin Scientific Supplier No. 23801008
Made with polypropylene, the tube rack can store up to 80 1.5 ml and 2.0 ml tubes, including its screw cap tubes. The rack can conveniently endure autoclaving and freezing temperatures of up to (-90C). This product measures 9" x 2 5/8" x 1 1/8" and is sold in a pack of 7 racks in assorted colors of blue, green, orange, red, yellow, violet, and natural.