Learn More
Andwin Scientific 80-PLACE TUBE RACKS FOR 0.5 1
SDP
Supplier:  Andwin Scientific 23801048
Encompass_Preferred
Made with polypropylene, the tube rack can store up to 80 1.5 ml and 2.0 ml tubes, including its screw cap tubes. The rack can conveniently endure autoclaving and freezing temperatures of up to (-90C). This product measures 9" x 2 5/8" x 1 1/8" and is sold in a pack of 7 racks in assorted neon colors of blue, green, orange, pink, yellow, black, and natural.
Catalog No. 50-819-984