Learn More
Organomation 80 position MULTIVAP Parallel Evaporator
SDP
Supplier:  Organomation 11880
Encompass_Preferred
The 80 position MULTIVAP parallel evaporator is a high temperature dry block model. This instrument contains a rectangular gas distribution system mounted on a frame and dual post assembly. The manifold delivers nitrogen gas to the needles or glass pipettes, which direct the gas onto the sample’s surface. This method results in rapid solvent evaporation and sample concentration. In order to conserve nitrogen gas, the manifold has a toggle switch for every row of 8 samples, which allows individual rows to be shut off when evaporating batches of less than 80 samples. Nitrogen gas flow is also controlled by an adjustable flow meter and a built-in pressure regulator. The 80 position MULTIVAP small vial evaporator uses a heated aluminum dry block to reach evaporative temperatures ranging from 30°C to 120°C. Sample blocks are customized, within a range of 10-17 mm, for the user’s sample size for maximum and even heat transfer.
Catalog No. NC2301752
Add to Cart
Request a Quote