Learn More
Sigma Aldrich Fine Chemicals Biosciences AxySeal sealing film with acrylic adhesive, for ELISA assays and T/C plates size 80 mum, polyester, pkg of 5x100sheets/cs, sterile |
Supplier:  Sigma Aldrich Fine Chemicals Biosciences AXYPCRSPS
Encompass
AxySeal sealing film with acrylic adhesive, for ELISA assays and T/C plates size 80 mum, polyester, pkg of 5x100sheets/cs, sterile |
Catalog No. NC2308348