Learn More
Teknova 80% (v/v) Glycerol Solution. 250mL, Sterile. 8 pack.
SDP
Catalog No. 502745922
Encompass
Glycerol solution is a versatile solvent that can be used in a variety of applications including electrophoresis, bacterial stock cultures, as an additive as a carbon source, and stabilizer.
Catalog No. 50-274-5922 Supplier Teknova Supplier No. G87978PK
Glycerol solution is a versatile solvent that can be used in a variety of applications including electrophoresis, bacterial stock cultures, as an additive as a carbon source, and stabilizer.