Learn More
Hamilton™ 800 Microliter Syringes, No Needle Included
Catalog No. 14824672 Shop All Hamilton Company Products
Catalog No. 14-824-672 Supplier Hamilton™ Supplier No. 764501
Add to Cart
Add to Cart
Attached to the flange of the 700 series syringe is a blue Syringe Holder that provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies. Additionally it provides support to small volume plungers that are prone to bending during injection. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 250 μL.
  • Ideal for dispensing volumes from 0.5 μL up to 250 μL
  • A variety of different terminations and needle options are available
  • Syringe Holder is attached to the flange that prevents the barrel from heating up and provides support to small volume plungers that are prone to bending