Learn More
Hamilton™ 800 Microliter Syringes, No Needle Included
Catalog No. 14824673 Shop All Hamilton Company Products
Catalog No. 14-824-673 Supplier Hamilton™ Supplier No. 764601
Add to Cart
Add to Cart
Attached to the flange of the 700 series syringe is a blue Syringe Holder that provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies. Additionally it provides support to small volume plungers that are prone to bending during injection. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 250 μL.
  • Ideal for dispensing volumes from 0.5 µL up to 10 µL
  • Syringe holder is permanently attached to the syringe barrel
Specifications
Material Borosilicate Glass
Needle Gauge No Needle Included
Product Type Microliter Syringe
Connection Type Removable
Graduated Yes