Learn More
Moldex™ 8000 Series Half-Mask Respirators Cartridges
SDP
Catalog No. 18102 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Cartridge Type:
Organic Vapor Cartridge
Organic Vapor, Acid Gas Cartridge
Particulate
For Use With (Application):
For protection against asbestos, lead, inorganic arsenic, cadmium, for use in environments where oil based particulates are present
For protection against organic vapors
For protection against organic vapors, chlorine, hydrogen chloride, sulfur dioxide, chlorine dioxide, hydrogen sulfide and hydrogen fluoride
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type For Use With (Application)
18-102 Organic Vapor Cartridge For protection against organic vapors
18-104 Organic Vapor, Acid Gas Cartridge For protection against organic vapors, chlorine, hydrogen chloride, sulfur dioxide, chlorine dioxide, hydrogen sulfide and hydrogen fluoride
19-003-347 Particulate For protection against asbestos, lead, inorganic arsenic, cadmium, for use in environments where oil based particulates are present
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-102 Supplier Moldex™ Supplier No. 8100
This item is not returnable. View return policy
For use with 8000 series half mask respirators
  • Use with 8910 N95, 8970 R95 or 8940 P100 filters against particulates
  • The P100 cartridge can be used for both oil and non-oil based particulates
  • 100% PVC-Free™
Specifications
Cartridge Type Organic Vapor Cartridge
Certifications/Compliance NIOSH
Product Type Organic Vapor Cartridge
For Use With (Application) For protection against organic vapors
For Use With (Equipment) Series 8000 half mask, n95, r95, p100 filter
Quantity 10/Pk.