Description
- Use with 8910 N95, 8970 R95 or 8940 P100 filters against particulates
- The P100 cartridge can be used for both oil and non-oil based particulates
- 100% PVC-Free™
Specifications
Specifications
| Cartridge Type | Particulate |
| Certifications/Compliance | NIOSH |
| Filter Type | P100 |
| Product Type | P100 Particulate Respirator Filter |
| For Use With (Application) | For protection against asbestos, lead, inorganic arsenic, cadmium, for use in environments where oil based particulates are present |
| For Use With (Equipment) | Series 8000 half mask, p100, r95 or n95 particulate filter |
| Quantity | 10/Pk. |