Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ 8000 Series Paint Spray/Pesticide Pre-Assembled Respirators
SDP

Catalog No. 19003343B Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-003-343B Medium
19-003-343A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-003-343B Supplier Moldex™ Supplier No. 8112N

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pre-assembled half-mask respirators equipped for oil-free paint spray and pesticide work environments. Kit includes 8000 Series facepiece, 8100 Organic Vapor Cartridges, 8910 N95 Pre-Filters and 8020 Pre-Filter Retainers.

  • 8000 Series half-mask pre-assembled with OV cartridges, N95 pre-filters and pre-filter retainers
  • For oil-free paint sprays and pesticides
  • Soft, durable facepiece and lightweight design with extra-wide sealing
  • Exhalation valve positioned for better communication
  • Cartridges have a snap-in mount for easy replacement and no misthreading
  • Living hinge allows head harness to be folded comfortably around neck when not in use
  • Low profile, compact design fits well under welding helmets and with safety glasses
  • Maintenance made simple with very few replacement parts

1 Includes two 19-003-346 N95 particulate prefilters, two 18-110 retainers, and two 18-102 organic vapor cartridges.

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84, TC-23C-1223
Color Gray
Filter Type N95
Material Thermoplastic Elastomer
Product Type Pre-Assembled Half-Mask Respirator
Size Medium
For Use With (Application) Paint spray and pesticide applications which are oil-free
Includes 8000 Facepiece, 8100 Organic Vapor Cartridges, 8910 N95 Pre-Filters and 8020 Pre-Filter Retainers
Model 8112N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.