Learn More
LICOR IRDye 800CW DBCO Infrared Dye (0.5 mg)
Supplier:  LICOR 92950000
Encompass_Preferred
IRDye 800CW DBCO provides the functionality for preferential labeling of molecules that contain azide groups (–N3) with IRDye 800CW.
Catalog No. NC0902760