Learn More
Ace Glass, Inc. Combo mantle, 3 position, low temperature, fits 800mL round bottom flasks, max diameter 4.56 inches, 240v 225 watts per position, order code (-63) or (-73) power supply separately
SDP
Catalog No. 50983294
Encompass
Combo mantle, 3 position, low temperature, fits 800mL round bottom flasks, max diameter 4.56 inches, 240v 225 watts per position, order code (-63) or (-73) power supply separately
Catalog No. 50-983-294 Supplier Ace Glass, Inc. Supplier No. 12061137
Combo mantle, 3 position, low temperature, fits 800mL round bottom flasks, max diameter 4.56 inches, 240v 225 watts per position, order code (-63) or (-73) power supply separately