Learn More
CELLTREAT Scientific Products 800mL Suspension Culture Flask - Vent Cap, Sterile, Case Quantity 25
SDP
Catalog No. 5000670044
Encompass_Preferred
Stable base enables filling with flask standing upright
Convenient zip-closure bag is a practical alternative to folding and taping opened bags
Vent caps feature 0.22μm hydrophobic membrane
Large opening, canted neck design and moderate angles allow easy access to entire flask surface
Molded, interlocking T design for improved stacking - included on 75, 182 & 300cm² and 250, 600 & 850mL size flasks only
Flask is printed with lot number and expiration date for easy identification and tracking
150cm2 Flask features U-shaped design for more efficient cell harvesting
Catalog No. 50-006-70044 Supplier CELLTREAT Scientific Products Supplier No. 229560
Stable base enables filling with flask standing upright
Convenient zip-closure bag is a practical alternative to folding and taping opened bags
Vent caps feature 0.22μm hydrophobic membrane
Large opening, canted neck design and moderate angles allow easy access to entire flask surface
Molded, interlocking T design for improved stacking - included on 75, 182 & 300cm² and 250, 600 & 850mL size flasks only
Flask is printed with lot number and expiration date for easy identification and tracking
150cm2 Flask features U-shaped design for more efficient cell harvesting