Learn More
Sino Biological Recombinant Rat IL-2R beta/CD122 Protein (His Tag)
Supplier:  Sino Biological 80340R08H100
Encompass_Preferred
A DNA sequence encoding the rat IL2RB (NP_037327.1) (Met1-Glu239) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-202-1287