Learn More
Metrohm USA 807 DOSING UNIT 20ML
SDP
Catalog No. NC0270616 Shop All Metrohm USA Products
Encompass
Dosing unit with integrated data chip with 20 mL glass cylinder and light protection, mountable on reagent bottle with ISO/DIN GL45 thread. FEP tubing connection, anti-diffusion buret tip
This item is not returnable. View return policy
Catalog No. NC0270616 Supplier Metrohm USA Supplier No. 63032220
Dosing unit with integrated data chip with 20 mL glass cylinder and light protection, mountable on reagent bottle with ISO/DIN GL45 thread. FEP tubing connection, anti-diffusion buret tip