Learn More
Metrohm USA 807 Dosing Unit ETFE 50 mL for dosing alkali reagents or hydrofluoric acid (HF) with Dosinos. Less precision and reproducibility is to be expected in comparison with glass cylinders.
SDP
Catalog No. NC3718581 Shop All Metrohm USA Products
Encompass
Material- ETFE,Material remark- Cylinder, Material 2- PVDF, Material remark 2- Housing, Volume (mL) 50
Catalog No. NC3718581 Supplier Metrohm USA Supplier No. 6.1575.250
Material- ETFE,Material remark- Cylinder, Material 2- PVDF, Material remark 2- Housing, Volume (mL) 50