Learn More
MSC Ferraz Shawmut TRS80R 600 VAC/VDC, 80 Amp, Time Delay General Purpose Fuse Clip Mount, 7-7/8" Overall Length, 100 at DC, 200 at AC kA Rating, 1-5/16" Diam
Catalog No. 501299872
Encompass
Ferraz Shawmut TRS80R 600 VAC/VDC, 80 Amp, Time Delay General Purpose Fuse Clip Mount, 7-7/8" Overall Length, 100 at DC, 200 at AC kA Rating, 1-5/16" Diam
Catalog No. 50-129-9872 Supplier MSC Supplier No. 54006549
Ferraz Shawmut TRS80R 600 VAC/VDC, 80 Amp, Time Delay General Purpose Fuse Clip Mount, 7-7/8" Overall Length, 100 at DC, 200 at AC kA Rating, 1-5/16" Diam