Learn More
Chemglass Life Sciences 80mm Ground Flat Flange, Tube ID: 75mm, Tube OD: 80mm
SDP
Catalog No. CG13503
Encompass_Preferred
Mating end of flange has a ground surface to provide for a liquid tight seal when greased and clamped with the CG-139 horseshoe clamp. 80mm Ground Flat Flange, Tube ID: 75mm, Tube OD: 80mm.
Catalog No. CG13503 Supplier Chemglass Life Sciences Supplier No. CG13503
Mating end of flange has a ground surface to provide for a liquid tight seal when greased and clamped with the CG-139 horseshoe clamp. 80mm Ground Flat Flange, Tube ID: 75mm, Tube OD: 80mm.