Learn More
Chemglass Life Sciences Clamp, Horseshoe Style, 80mm Flange
SDP
Catalog No. CG13903
Encompass_Preferred
Clamp, Horseshoe Style, 80mm Flange. Horseshoe shaped clamp used to connect CG-138 flat o-ring flange. Design allows clamp to be installed or removed easily. Made of cast aluminum with chemically resistant green epoxy coating. Three adjustable tension locking screws assure even clamping pressure.
Catalog No. CG13903 Supplier Chemglass Life Sciences Supplier No. CG13903
Clamp, Horseshoe Style, 80mm Flange. Horseshoe shaped clamp used to connect CG-138 flat o-ring flange. Design allows clamp to be installed or removed easily. Made of cast aluminum with chemically resistant green epoxy coating. Three adjustable tension locking screws assure even clamping pressure.