Learn More
Ace Glass, Inc. 80mm adapter, nylon, polyethylene packing slip bottom drip
SDP
Catalog No. 50978487
Encompass_Preferred
11mm bottom-drip, Luer-Lok, PTFE adapter for use with all #11 Ace- Thred chromatography columns. Bore is 1mm. Supplied with one FETFE O-Ring and one replaceable polyethylene packing support.
Catalog No. 50-978-487 Supplier Ace Glass, Inc. Supplier No. 583727
11mm bottom-drip, Luer-Lok, PTFE adapter for use with all #11 Ace- Thred chromatography columns. Bore is 1mm. Supplied with one FETFE O-Ring and one replaceable polyethylene packing support.