Learn More
Chemglass Life Sciences 80mm O-Ring Flat Flange, Tube ID: 75mm, Tube OD: 80mm, O-Ring Size 339
SDP
Catalog No. CG13803
Encompass_Preferred
80mm O-Ring Flat Flange, Tube ID: 75mm, Tube OD: 80mm, O-Ring Size 339. Similar to CG-135 flange but with the mating end of the flange having a precision formed o-ring groove instead of being ground. Joint provides a grease-free vacuum tight seal when used with the CG-139 clamp. O-ring is included with each flange and can be ordered separately, see CG-305.
Catalog No. CG13803 Supplier Chemglass Life Sciences Supplier No. CG13803
80mm O-Ring Flat Flange, Tube ID: 75mm, Tube OD: 80mm, O-Ring Size 339. Similar to CG-135 flange but with the mating end of the flange having a precision formed o-ring groove instead of being ground. Joint provides a grease-free vacuum tight seal when used with the CG-139 clamp. O-ring is included with each flange and can be ordered separately, see CG-305.