Learn More
Nanocomposix BioReady Gold Nanospheres - Carboxyl - 80 nm, 20 OD in water, 30 mL
SDP
Supplier:  Nanocomposix AUXR8030M
Encompass_Preferred
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.80 nm spherical gold nanoparticles for covalent conjugation combine strong plasmonic optical properties with readily biofunctionalized surface chemistry. These nanoparticles are used in applications such as immunodiagnostics, lateral flow assays, biosensors, and more.BioReady Gold Nanospheres have carboxyl covalently bound to the metal surface. Antibodies and other amine-containing biomolecules efficiently bind to carboxyl gold nanopheres using EDC-NHS chemistry to make highly stable conjugates. Review our conjugation protocol or contact our experts for support.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.
Catalog No. NC2885655