Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ 80nm Colloidal Gold Nanoparticles 1OD
SDP

Catalog No. 38010100
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-010-100 100 mL
38-010-010 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-010-100 Supplier Novus Biologicals™ Supplier No. 38010100

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Ultra high quality with narrow size distribution and uniform shape

80 nm colloidal gold is an aqueous suspension of spherical metallic nanoparticles manufactured to the highest standard to ensure:
  • Uniform spherical shape
  • Narrow size distribution: 80 nm ± 8nm, <10% CV
  • Ultra high quality
  • Batch-to-batch consistency
  • High efficiency antibody binding for maximum sensitivity
  • Available at high concentrations to simplify assay development

Specifications

Quantity 100 mL
Content And Storage 4°C Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.