Learn More
Perkin Elmer US LLC 80Spec Grazing Angle Specular Reflectance Accessory
Supplier:  Perkin Elmer US LLC L1272248
Encompass_Preferred
The 80Spec accessory specializes in the specular reflectance measurements of thin films and mono-molecular layers. Samples are laid face down across the top of the accessory and the spectrum of the film is measured. This accessory is compatible with Spectrum One/100/400/65, Spectrum Two, and Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-3607