Learn More
Andwin Scientific 80WELL MICROTUBE RACK 5 x 16 in, 5PK
SDP
Catalog No. 50727581
Encompass_Preferred
Andwin Scientific 80WELL MICROTUBE RACK is made of polypropylene and is autoclavable. This product's conical bottoms hold 1.5-2.0mL microtubes in a 5 x 16 in. array. This product is sold in a pack of 5 in assorted colors.
Catalog No. 50-727-581 Supplier Andwin Scientific Supplier No. 61603A
Andwin Scientific 80WELL MICROTUBE RACK is made of polypropylene and is autoclavable. This product's conical bottoms hold 1.5-2.0mL microtubes in a 5 x 16 in. array. This product is sold in a pack of 5 in assorted colors.