Learn More
Analytical Sales & Services 81 Position Storage Box for Large 3-5mL Sample Tubes
Catalog No. 5000804554
Encompass
Advantage™ 81 Position Storage Box for Large 3-5mL Sample Tubes\n\n• Made of high impact polystyrene.\n• Temperature range: -80ºC to 80ºC. Not autoclavable.\n• Color: Blue\n\nQty/Pkg: 10
Catalog No. 50-008-04554 Supplier Analytical Sales & Services Supplier No. 96581B
Advantage™ 81 Position Storage Box for Large 3-5mL Sample Tubes\n\n• Made of high impact polystyrene.\n• Temperature range: -80ºC to 80ºC. Not autoclavable.\n• Color: Blue\n\nQty/Pkg: 10