Learn More
Sigma Aldrich Fine Chemicals Biosciences 81-Well microtube storage
Catalog No. 502900064
Encompass
Compact footprint for microtube and cryogenic vial storage.Made from polypropyleneStore microtubes or cryogenic vials Easy open friction fit lidSafe for low temperature storageBlack light tight version available for light sensitive samplesTubes 1.5/2.0 mLWells 81Well Dia 12.3 mmDimensions 5.1 x 5.1 x 1.9 in (13 x 13 x 4.7 cm)AutoclavableEach Heathrow Scientific Microtube Storage boxes is designed for quick visual identification.Uniform wells with location IDTube capacity is indicated on the outside of the boxTranslucent vibrant colors with a frosted labeling areaDebossed with a distinctive wave design
Catalog No. 50-290-0064 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS120040
Compact footprint for microtube and cryogenic vial storage.Made from polypropyleneStore microtubes or cryogenic vials Easy open friction fit lidSafe for low temperature storageBlack light tight version available for light sensitive samplesTubes 1.5/2.0 mLWells 81Well Dia 12.3 mmDimensions 5.1 x 5.1 x 1.9 in (13 x 13 x 4.7 cm)AutoclavableEach Heathrow Scientific Microtube Storage boxes is designed for quick visual identification.Uniform wells with location IDTube capacity is indicated on the outside of the boxTranslucent vibrant colors with a frosted labeling areaDebossed with a distinctive wave design