Learn More
Electron Microscopy Sciences Microtube 81-Well Storage Rack, Green
SDP
Catalog No. 5034128
Encompass_Preferred
Durable polypropylene rack fits in standard 5.25" square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials. Features molded grid lines on the lid, imprinted coordinates on the bottom of base, and a molded reference point on the lid. Rack measures 5.125 x 5.125 x 1.75".
Color: Green
Catalog No. 50-341-28 Supplier Electron Microscopy Sciences Supplier No. 61600G
Durable polypropylene rack fits in standard 5.25" square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials. Features molded grid lines on the lid, imprinted coordinates on the bottom of base, and a molded reference point on the lid. Rack measures 5.125 x 5.125 x 1.75".
Color: Green