Learn More
Electron Microscopy Sciences Microtube 81-Well Storage Rack, Orange
SDP
Catalog No. 5034131
Encompass_Preferred
Durable polypropylene rack fits in standard 5.25" square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials. Features molded grid lines on the lid, imprinted coordinates on the bottom of base, and a molded reference point on the lid. Rack measures 5.125 x 5.125 x 1.75".
Color: Orange
Catalog No. 50-341-31 Supplier Electron Microscopy Sciences Supplier No. 61600O
Durable polypropylene rack fits in standard 5.25" square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials. Features molded grid lines on the lid, imprinted coordinates on the bottom of base, and a molded reference point on the lid. Rack measures 5.125 x 5.125 x 1.75".
Color: Orange