Learn More
Biolegend APC/Fire™ 810 anti-human CD38
Supplier:  Biolegend 356643
Encompass_Preferred
APC/Fire™ 810 anti-human CD38 [HB-7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1853805