Learn More
eMolecules​ 81190-89-8 | Methyl 4-bromo-1H-pyrazole-3-carboxylate | Ambeed | MFCD00619124 | 205.011 | C5H5BrN2O2 | 95.000 | COC(=O)c1n[nH]cc1Br | 1g | 552727554
Catalog No. 5000488746 Shop All eMolecules Inc. Products
Encompass_Preferred
Methyl 4-bromo-1H-pyrazole-3-carboxylate | Ambeed | 81190-89-8 | MFCD00619124 | 205.011 | C5H5BrN2O2 | 95.000 | COC(=O)c1n[nH]cc1Br | 1g | 552727554
Catalog No. 50-004-88746 Supplier eMolecules​ Supplier No. 552727554
Methyl 4-bromo-1H-pyrazole-3-carboxylate | Ambeed | 81190-89-8 | MFCD00619124 | 205.011 | C5H5BrN2O2 | 95.000 | COC(=O)c1n[nH]cc1Br | 1g | 552727554