Learn More
Seca Corp 813 High capacity digital flat scale for individual patient use
SDP
Catalog No. NC9083247 Shop All Seca Corp Products
Encompass_Preferred
seca has designed the seca 813 with a high capacity of up to 440 lbs and an extra wide platform. The tough rubber mat ensures a secure stance and flat construction allows for easy accessibility. • Solid steel frame, up to 440 lbs capacity • Non-slip rubber base for safe standing during measuring • Large LCD figures for easy reading • Step-off function for weighing without switching on scale • Capacity 440 lbs / 200 kg • Graduation 0.2 lb / 100 g • Functions • Auto-HOLD, lbs/kg switch-over, Step-off, tap-on automatic switch-on, automatic switch-off
Catalog No. NC9083247 Supplier Seca Corp Supplier No. 8131321009
seca has designed the seca 813 with a high capacity of up to 440 lbs and an extra wide platform. The tough rubber mat ensures a secure stance and flat construction allows for easy accessibility. • Solid steel frame, up to 440 lbs capacity • Non-slip rubber base for safe standing during measuring • Large LCD figures for easy reading • Step-off function for weighing without switching on scale • Capacity 440 lbs / 200 kg • Graduation 0.2 lb / 100 g • Functions • Auto-HOLD, lbs/kg switch-over, Step-off, tap-on automatic switch-on, automatic switch-off