Learn More
eMolecules​ 819058-34-9 | (4-AMINO-3-FLUOROPHENYL)BORONIC ACID PINACOL ESTER | AstaTech | MFCD09033884 | 237.080 | C12H17BFNO2 | 95.000 | CC1(C)OB(OC1(C)C)c1ccc(N)c(F)c1 | 5g | 233630498
Catalog No. 5000163239 Shop All eMolecules Inc. Products
Encompass_Preferred
(4-AMINO-3-FLUOROPHENYL)BORONIC ACID PINACOL ESTER | AstaTech | 819058-34-9 | MFCD09033884 | 237.080 | C12H17BFNO2 | 95.000 | CC1(C)OB(OC1(C)C)c1ccc(N)c(F)c1 | 5g | 233630498
Catalog No. 50-001-63239 Supplier eMolecules​ Supplier No. 233630498
(4-AMINO-3-FLUOROPHENYL)BORONIC ACID PINACOL ESTER | AstaTech | 819058-34-9 | MFCD09033884 | 237.080 | C12H17BFNO2 | 95.000 | CC1(C)OB(OC1(C)C)c1ccc(N)c(F)c1 | 5g | 233630498