Learn More
Perkin Elmer US LLC 81-Position Sample Tray and Diluent Tank for Series 200 Autosampler
Catalog No. 502111761
Encompass_Preferred
PerkinElmer has researched, engineered, and developed the Series 200 Autosampler to ensure precision, flexibility, reliability, and ease-of-use. Our unique XYZ design allows you to take advantage of the built-in extra capabilities including dilution, derivatization, and standard addition all without additional hardware or cost.
Catalog No. 50-211-1761 Supplier Perkin Elmer US LLC Supplier No. N2930314
PerkinElmer has researched, engineered, and developed the Series 200 Autosampler to ensure precision, flexibility, reliability, and ease-of-use. Our unique XYZ design allows you to take advantage of the built-in extra capabilities including dilution, derivatization, and standard addition all without additional hardware or cost.